Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stard6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289648.1 | NP_001276577.1 | 6, 7 | 561 | ||
NM_001289649.1 | NP_001276578.1 | 6, 7 | 540 | |||
NM_001289650.1 | NP_001276579.1 | 6, 7 | 575 | |||
NM_029019.4 | NP_083295.1 | 6, 7 | 630 | |||
GenBank mRNA | AF480303.1 | - | 3, 4 | 277 | ||
AK005873.1 | - | 6, 7 | 575 | |||
AK014759.1 | - | 6, 7 | 541 | |||
AK016984.1 | - | 6, 7 | 541 | |||
BC061022.1 | - | 4, 5 | 313 | 6774873 | ||
BC087893.1 | - | 6, 7 | 552 | 6774832 | ||
JF440953.1 | - | 4, 5 | 349 | |||
JF440954.1 | - | 4, 5 | 349 |