Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168503.1 | NP_001161975.1 | 3 | 831 | ||
NM_130866.4 | NP_570936.1 | 4 | 1700 | |||
GenBank mRNA | AF378854.1 | - | 1 | 485 | ||
AK028467.1 | - | 4 | 1625 | |||
AK036356.1 | - | 3 | 832 | |||
BC120603.1 | - | 2 | 674 | 40129526 | ||
BC120629.1 | - | 2 | 674 | 40129552 |