Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Card14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130886.3 | NP_570956.1 | 6 | 1067 | ||
GenBank mRNA | AF363457.1 | - | 6 | 1046 | ||
AK028888.1 | - | 1 | 53 | |||
AK086176.1 | - | 7 | 1316 | |||
BC004692.1 | - | 3 | 502 | |||
BC029102.1 | - | 6 | 1067 | 3979883 |