Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn4ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130892.4 | NP_570962.2 | 3 | 915 | ||
GenBank mRNA | AF336862.1 | - | 3 | 645 | ||
AK050324.1 | - | 3 | 913 | |||
AK085858.1 | - | 3 | 895 | |||
AK088029.1 | - | 3 | 916 | |||
AK204568.1 | - | 3 | 245 | |||
BC024116.1 | - | 3 | 885 | 5100503 |