Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133216.3 | NP_573479.3 | 10 | 1102 | ||
GenBank mRNA | AF363970.1 | - | 10 | 1042 | ||
AK052094.1 | - | 2 | 244 | |||
AK146033.1 | - | 10 | 1082 | |||
AK149676.1 | - | 10 | 1041 | |||
AK158597.1 | - | 8 | 1060 | |||
AK166715.1 | - | 10 | 1002 | |||
AK169024.1 | - | 9 | 928 | |||
AK177494.1 | - | 1 | 88 | |||
AK185825.1 | - | 1 | 89 | |||
BC021534.1 | - | 1 | 37 | |||
BC065174.1 | - | 10 | 990 | 6415314 |