Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199198.2 | NP_954668.2 | 12 | 1316 | ||
GenBank mRNA | AK040235.1 | - | 12 | 1279 | ||
AK045028.1 | - | 11 | 1527 | |||
AK154045.1 | - | 11 | 1211 | |||
BC013700.1 | - | 12 | 1280 | |||
BC064018.1 | - | 12 | 1243 | 3980699 |