Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133241.2 | NP_573504.1 | 4, 5 | 375 | ||
GenBank mRNA | AF449425.1 | - | 4, 5 | 829 | ||
AK030396.1 | - | 4, 5 | 374 | |||
AK043757.1 | - | 4, 5 | 375 | |||
AK122190.1 | - | 4, 5 | 817 | |||
BC024719.1 | - | 4, 5 | 375 | 5363238 |