Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133241.2 | NP_573504.1 | 10, 11 | 949 | ||
GenBank mRNA | AF449425.1 | - | 10, 11 | 1403 | ||
AK030396.1 | - | 10, 11 | 948 | |||
AK043757.1 | - | 10, 11 | 949 | |||
AK122190.1 | - | 10, 11 | 1391 | |||
BC024719.1 | - | 10, 11 | 949 | 5363238 |