Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291114.1 | NP_001278043.1 | 17 | 1902 | ||
NM_001291115.1 | NP_001278044.1 | 17 | 1697 | |||
NM_133242.3 | NP_573505.2 | 17 | 1920 | |||
GenBank mRNA | AK134091.1 | - | 7 | 472 | ||
AK135289.1 | - | 16 | 1528 | |||
AK146046.1 | - | 17 | 1697 | |||
AK147076.1 | - | 17 | 1865 | |||
AK154989.1 | - | 17 | 1833 | |||
BC004000.1 | - | 13 | 1252 | |||
BC023205.1 | - | 17 | 1822 | |||
BC030493.1 | - | 15 | 1517 | 5400675 | ||
BC082607.1 | - | 15 | 1492 | 30667785 | ||
BC086645.1 | - | 17 | 1843 | 6817169 | ||
BC092268.1 | - | 15 | 1651 |