Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076676.2 | NP_001070144.1 | 24 | 2830 | ||
NM_001252486.1 | NP_001239415.1 | 24 | 2842 | |||
NM_133247.3 | NP_573510.2 | 24 | 2866 | |||
GenBank mRNA | AF383174.1 | - | 24 | 2855 | ||
AK042458.1 | - | 1948 | ||||
AK082251.1 | - | 24 | 2813 | |||
AK082943.1 | - | 24 | 2813 | |||
AK087467.1 | - | 24 | 2846 | |||
AK122442.1 | - | 21 | 5324 | |||
AK194905.1 | - | 2 | 117 | |||
BC005506.1 | - | 24 | 2795 | |||
BC031366.1 | - | 16 | 1982 | |||
BC036980.1 | - | 17 | 2042 | |||
BC049870.1 | - | 11 | 1107 | |||
BC067399.1 | - | 22 | 2605 | |||
BC089315.1 | - | 24 | 2788 |