Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161738.1 | NP_001155210.1 | 15 | 1448 | ||
NM_133248.2 | NP_573511.2 | 15 | 1462 | |||
GenBank mRNA | AJ566083.1 | - | 14 | 1330 | ||
AK034160.1 | - | 15 | 1449 | |||
AK165661.1 | - | 15 | 1433 | |||
AK169045.1 | - | 15 | 1434 | |||
BC003446.1 | - | 15 | 1398 | 2651614 |