Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277968.1 | NP_001264897.1 | 2 | 349 | ||
NM_008534.3 | NP_032560.2 | 3 | 679 | |||
GenBank mRNA | AB196804.1 | - | 3 | 592 | ||
AB196805.1 | - | 3 | 592 | |||
AB196806.1 | - | 3 | 592 | |||
AB196807.1 | - | 3 | 592 | |||
AF244130.1 | - | 3 | 679 | |||
AF244131.1 | - | 3 | 679 | |||
AK040306.1 | - | 3 | 630 | |||
AK042288.1 | - | 3 | 618 | |||
AK087345.1 | - | 3 | 599 | |||
AK088815.1 | - | 2 | 298 | |||
AK134106.1 | - | 3 | 696 | |||
AK150121.1 | - | 3 | 606 | |||
BC055380.1 | - | 3 | 579 | |||
BC066212.1 | - | 3 | 567 | |||
BC095921.1 | - | 3 | 602 | 4935844 | ||
M84412.1 | - | 3 | 543 |