Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134159.4 | NP_598920.3 | 10, 11 | 1203 | ||
NM_178942.1 | NP_849273.1 | 11, 12 | 1275 | |||
GenBank mRNA | AF458066.1 | - | 10, 11 | 1084 | ||
AI463827.1 | - | 1, 2 | 188 | |||
AK075634.1 | - | 7, 8 | 693 | |||
BC004759.1 | - | 11, 12 | 1223 | 3154616 |