Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17rc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134159.4 | NP_598920.3 | 17 | 2185 | ||
NM_178942.1 | NP_849273.1 | 18 | 2257 | |||
GenBank mRNA | AF458066.1 | - | 17 | 2066 | ||
AK033890.1 | - | 1 | 108 | |||
AK040950.1 | - | 1 | 110 | |||
AK075634.1 | - | 14 | 1675 | |||
AW045329.1 | - | 1 | 101 | |||
BC004759.1 | - | 18 | 1913 | 3154616 |