Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012517.5 | NP_001012535.1 | 4 | 1534 | ||
NM_001286422.1 | NP_001273351.1 | 5 | 1639 | |||
NM_001286424.1 | NP_001273353.1 | 4 | 1427 | |||
NM_134161.3 | NP_598922.1 | 4 | 1534 | |||
GenBank mRNA | AJ880009.1 | - | 3 | 1147 | ||
AK147136.1 | - | 5 | 1595 | |||
AK169153.1 | - | 4 | 1488 | |||
BC022579.1 | - | 1 | 632 | |||
BC030474.1 | - | 4 | 1431 | 5368333 | ||
BC062113.1 | - | 4 | 1533 | 6816067 |