Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134420.4 | NP_599252.2 | 2597 | |||
GenBank mRNA | AB099881.1 | - | 1758 | |||
AF248494.1 | - | 1943 | ||||
AK029238.1 | - | 3519 | ||||
AK046993.1 | - | 3857 | ||||
AK053088.1 | - | 2248 | ||||
AK163670.1 | - | 2598 | ||||
AK163671.1 | - | 2598 | ||||
AY032863.1 | - | 1902 | ||||
AY049076.1 | - | 1857 | ||||
BC028856.1 | - | 1921 | 4165725 | |||
CN692543.1 | - | 417 |