Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nme7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293313.1 | NP_001280242.1 | 7 | 767 | ||
NM_138314.4 | NP_612187.2 | 7 | 770 | |||
NM_178071.6 | NP_835172.1 | 7 | 767 | |||
GenBank mRNA | AF202048.1 | - | 6 | 671 | ||
AK044962.1 | - | 7 | 741 | |||
AK085222.1 | - | 7 | 715 | |||
AK133221.1 | - | 1238 | ||||
AK144915.1 | - | 7 | 771 | |||
AK164652.1 | - | 7 | 715 | |||
BC038021.1 | - | 8 | 797 | 3963844 | ||
BC049225.1 | - | 6 | 698 | 3483725 |