Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008549.2 | NP_032575.2 | 7 | 1165 | ||
GenBank mRNA | AK144763.1 | - | 2080 | |||
BC138371.1 | - | 7 | 1150 | 8861391 | ||
BC138372.1 | - | 7 | 1150 | 8861392 | ||
X61172.1 | - | 7 | 1165 |