Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008550.2 | NP_032576.2 | 10 | 1520 | ||
GenBank mRNA | AB006458.1 | - | 10 | 1498 | ||
AK004900.1 | - | 10 | 1474 | |||
AK040388.1 | - | 10 | 1481 | |||
AK075649.1 | - | 2 | 185 | |||
AK165376.1 | - | 10 | 1513 | |||
BC066211.1 | - | 10 | 1471 | 30101816 |