Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008550.2 | NP_032576.2 | 11 | 1902 | ||
GenBank mRNA | AB006458.1 | - | 11 | 1880 | ||
AK004900.1 | - | 11 | 1856 | |||
AK040388.1 | - | 11 | 1863 | |||
AK075649.1 | - | 3 | 567 | |||
AK165376.1 | - | 11 | 1895 | |||
BC066211.1 | - | 11 | 1853 | 30101816 |