Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Maz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010772.1 | NP_034902.1 | 3, 4 | 1168 | ||
GenBank mRNA | AK209702.1 | - | 1, 2 | 19 | ||
BC004758.1 | - | 5, 6 | 705 | |||
DQ150586.1 | - | 2, 3 | 882 | |||
L04649.1 | - | 3, 4 | 1141 |