Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013594.2 | NP_038622.2 | 3 | 382 | ||
GenBank mRNA | AF072240.1 | - | 3 | 385 | ||
AK004624.2 | - | 3 | 383 | |||
AK156401.1 | - | 3 | 397 | |||
AK162858.1 | - | 3 | 382 | |||
AK204682.1 | - | 1 | 71 | |||
BC069837.1 | - | 3 | 398 | 30435760 |