Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcpt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008572.1 | NP_032598.1 | 4 | 586 | ||
GenBank mRNA | AK136485.1 | - | 4 | 606 | ||
BC119342.1 | - | 4 | 662 | 8734091 | ||
BC120607.1 | - | 4 | 659 | 40129530 | ||
BC144934.1 | - | 4 | 748 | |||
X78545.1 | - | 4 | 586 |