Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161413.1 | NP_001154885.1 | 3 | 953 | ||
NM_008577.4 | NP_032603.3 | 2 | 712 | |||
GenBank mRNA | AB023408.1 | - | 2 | 667 | ||
AK090046.1 | - | 2 | 690 | |||
AK156285.1 | - | 2 | 184 | |||
AK161280.1 | - | 3 | 954 | |||
AK165417.1 | - | 2 | 694 | |||
AK165550.1 | - | 2 | 690 | |||
AK180963.1 | - | 1 | 171 | |||
AK183775.1 | - | 1 | 77 | |||
AK215373.1 | - | 2 | 178 | |||
BC065173.1 | - | 2 | 672 | 6415269 | ||
U25708.1 | - | 2 | 689 | |||
X14309.1 | - | 2 | 654 |