Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 16 | 1688 | ||
GenBank mRNA | AK010498.1 | - | 16 | 1654 | ||
AK011932.1 | - | 16 | 1688 | |||
AK129076.1 | - | 16 | 1641 | |||
AK137000.1 | - | 16 | 1714 | |||
AK145021.1 | - | 16 | 1623 | |||
AK145316.1 | - | 16 | 1597 | |||
AK161029.1 | - | 16 | 1697 | |||
AK164138.1 | - | 16 | 1655 | |||
AK167457.1 | - | 16 | 1612 | |||
BC020416.1 | - | 15 | 1523 | |||
BC085276.1 | - | 16 | 1721 | 30443236 | ||
L76158.1 | - | 16 | 1619 | |||
X95351.1 | - | 16 | 1626 |