Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Melk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010790.2 | NP_034920.2 | 14 | 1412 | ||
GenBank mRNA | AK010498.1 | - | 14 | 1378 | ||
AK011932.1 | - | 14 | 1412 | |||
AK129076.1 | - | 14 | 1365 | |||
AK137000.1 | - | 14 | 1438 | |||
AK145021.1 | - | 14 | 1347 | |||
AK145316.1 | - | 14 | 1321 | |||
AK161029.1 | - | 14 | 1421 | |||
AK164138.1 | - | 14 | 1379 | |||
AK167457.1 | - | 14 | 1336 | |||
BC020416.1 | - | 13 | 1247 | |||
BC085276.1 | - | 14 | 1445 | 30443236 | ||
L76158.1 | - | 14 | 1343 | |||
X95351.1 | - | 14 | 1350 |