Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110253.2 | NP_001103723.2 | 13 | 3792 | ||
NM_148925.3 | NP_683727.3 | 14 | 4039 | |||
GenBank mRNA | AJ428065.1 | - | 14 | 3944 | ||
AK044587.1 | - | 6 | 1101 | |||
BC022708.1 | - | 1 | 44 | |||
CB244718.1 | - | 5 | 625 |