Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fyco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110253.2 | NP_001103723.2 | 4 | 483 | ||
NM_148925.3 | NP_683727.3 | 5 | 730 | |||
GenBank mRNA | AJ428065.1 | - | 5 | 635 | ||
AK013060.1 | - | 4 | 471 | |||
AK081259.1 | - | 4 | 466 | |||
BC053712.1 | - | 3 | 391 | |||
CA467222.1 | - | 4 | 505 |