Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252292.1 | NP_001239221.1 | 7 | 871 | ||
NM_001252293.1 | NP_001239222.1 | 6 | 592 | |||
NM_008590.2 | NP_032616.1 | 6 | 770 | |||
GenBank mRNA | AK032881.1 | - | 6 | 769 | ||
AK034949.1 | - | 6 | 768 | |||
AK146457.1 | - | 7 | 871 | |||
AK168714.1 | - | 6 | 768 | |||
AK168732.1 | - | 6 | 768 | |||
AK168743.1 | - | 6 | 768 | |||
AK168995.1 | - | 6 | 768 | |||
AK169095.1 | - | 6 | 768 | |||
AK169266.1 | - | 6 | 768 | |||
BC006639.1 | - | 6 | 753 | 3482928 | ||
CF916557.1 | - | 6 | 584 | |||
D16262.1 | - | 6 | 757 |