Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164167.1 | NP_001157639.1 | 2 | 619 | ||
NM_001164168.1 | NP_001157640.1 | 2 | 635 | |||
NM_001164169.1 | NP_001157641.1 | 2 | 619 | |||
NM_001164170.1 | NP_001157642.1 | 2 | 635 | |||
NM_008602.4 | NP_032628.3 | 2 | 619 | |||
GenBank mRNA | AF039567.1 | - | 1 | 64 | ||
AK014871.1 | - | 2 | 610 | |||
AK029716.1 | - | 2 | 620 | |||
AK078813.1 | - | 2 | 589 | |||
AK086653.1 | - | 2 | 551 | |||
BC005596.1 | - | 2 | 496 | 3969546 | ||
BC034711.1 | - | 2 | 625 | 2631738 |