Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mme | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289462.1 | NP_001276391.1 | 10 | 1340 | ||
NM_001289463.1 | NP_001276392.1 | 10 | 1124 | |||
NM_008604.4 | NP_032630.2 | 10 | 974 | |||
GenBank mRNA | AK031446.1 | - | 10 | 975 | ||
AK033824.1 | - | 10 | 1035 | |||
AK040730.1 | - | 10 | 1012 | |||
AK052088.1 | - | 9 | 861 | |||
AK076908.1 | - | 10 | 1188 | |||
BC034092.1 | - | 10 | 996 | 4483558 | ||
BC066840.1 | - | 9 | 883 | |||
M81591.1 | - | 9 | 886 |