Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008605.3 | NP_032631.3 | 3, 4 | 569 | ||
GenBank mRNA | AK089295.1 | - | 538 | |||
AK089309.1 | - | 3, 4 | 538 | |||
AK089335.1 | - | 3, 4 | 534 | |||
AK089452.1 | - | 3, 4 | 538 | |||
AK089509.1 | - | 4, 5 | 412 | |||
AK089523.1 | - | 3, 4 | 538 | |||
AK090051.1 | - | 3, 4 | 540 | |||
AK150596.1 | - | 3, 4 | 538 | |||
AK151021.1 | - | 3, 4 | 538 | |||
AK151273.1 | - | 3, 4 | 538 | |||
AK151933.1 | - | 3, 4 | 538 | |||
AK155139.1 | - | 3, 4 | 537 | |||
AK155743.1 | - | 3, 4 | 538 | |||
AK155747.1 | - | 3, 4 | 538 | |||
AK155748.1 | - | 3, 4 | 538 | |||
AK155841.1 | - | 3, 4 | 596 | |||
AK155865.1 | - | 3, 4 | 537 | |||
AK170360.1 | - | 3, 4 | 538 | |||
AK170364.1 | - | 3, 4 | 538 | |||
AK170500.1 | - | 4, 5 | 412 | |||
AK170667.1 | - | 2, 3 | 1585 | |||
AK170798.1 | - | 3, 4 | 537 | |||
AK170804.1 | - | 3, 4 | 537 | |||
AK170976.1 | - | 3, 4 | 538 | |||
AK171006.1 | - | 3, 4 | 538 | |||
AK171036.1 | - | 3, 4 | 538 | |||
AK171176.1 | - | 3, 4 | 538 | |||
AK171428.1 | - | 3, 4 | 538 | |||
AK171435.1 | - | 2, 3 | 1585 | |||
AK171447.1 | - | 3, 4 | 538 | |||
AK171448.1 | - | 3, 4 | 538 | |||
AK171449.1 | - | 3, 4 | 538 | |||
AK171450.1 | - | 3, 4 | 538 | |||
AK171466.1 | - | 3, 4 | 538 | |||
AK171468.1 | - | 3, 4 | 538 | |||
AK171470.1 | - | 3, 4 | 538 | |||
AK171472.1 | - | 3, 4 | 538 | |||
AK171485.1 | - | 3, 4 | 538 | |||
AK171518.1 | - | 3, 4 | 538 | |||
AK171542.1 | - | 3, 4 | 538 | |||
AK171570.1 | - | 3, 4 | 538 | |||
BC019135.1 | - | 3, 4 | 532 | 5037674 | ||
CT010377.1 | - | 3, 4 | 485 | |||
M82831.1 | - | 3, 4 | 485 |