Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008608.3 | NP_032634.3 | 3 | 545 | ||
GenBank mRNA | AK040968.1 | - | 517 | |||
AK088476.1 | - | 3 | 545 | |||
AK134340.1 | - | 3 | 476 | |||
AK138611.1 | - | 3 | 486 | |||
AK149636.1 | - | 3 | 474 | |||
AK149907.1 | - | 3 | 525 | |||
AK149988.1 | - | 3 | 445 | |||
AK155117.1 | - | 3 | 547 | |||
AK163601.1 | - | 3 | 537 | |||
AK165014.1 | - | 3 | 546 | |||
BC076638.1 | - | 3 | 537 | 30658833 | ||
DQ249870.1 | - | 3 | 283 | |||
U54984.1 | - | 3 | 529 | |||
X83536.2 | - | 3 | 455 |