Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008608.3 | NP_032634.3 | 8, 9 | 1567 | ||
GenBank mRNA | AK040968.1 | - | 1540 | |||
AK088476.1 | - | 8, 9 | 1567 | |||
AK134340.1 | - | 8, 9 | 1499 | |||
AK138611.1 | - | 8, 9 | 1508 | |||
AK149636.1 | - | 8, 9 | 1496 | |||
AK149907.1 | - | 8, 9 | 1547 | |||
AK149988.1 | - | 8, 9 | 1467 | |||
AK155117.1 | - | 8, 9 | 1569 | |||
AK163601.1 | - | 8, 9 | 1559 | |||
AK165014.1 | - | 8, 9 | 1568 | |||
AK185366.1 | - | 1, 2 | 115 | |||
AK189497.1 | - | 1, 2 | 115 | |||
BC076638.1 | - | 8, 9 | 1559 | 30658833 | ||
DQ249870.1 | - | 8, 9 | 1305 | |||
U54984.1 | - | 8, 9 | 1551 | |||
X83536.2 | - | 8, 9 | 1477 |