Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019724.3 | NP_062698.2 | 3 | 578 | ||
GenBank mRNA | AB021228.1 | - | 3 | 393 | ||
AK046170.1 | - | 3 | 437 | |||
AK048543.1 | - | 3 | 458 | |||
AK053598.1 | - | 3 | 468 | |||
AK077939.1 | - | 622 | ||||
BC057926.1 | - | 3 | 580 | 5292478 |