Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289442.1 | NP_001276371.1 | 4 | 434 | ||
NM_010815.3 | NP_034945.1 | 3 | 529 | |||
GenBank mRNA | AF053405.1 | - | 3 | 497 | ||
AF055465.1 | - | 3 | 529 | |||
AF129477.1 | - | 3 | 492 | |||
AF236118.1 | - | 3 | 499 | |||
AJ011735.1 | - | 3 | 495 | |||
AK156939.1 | - | 3 | 508 | |||
BC034859.1 | - | 4 | 671 | |||
BC052496.1 | - | 4 | 446 | 1446918 |