Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010817.2 | NP_034947.1 | 7 | 709 | ||
GenBank mRNA | AK031013.1 | - | 7 | 660 | ||
AK146128.1 | - | 1434 | ||||
AK159825.1 | - | 7 | 661 | |||
BC013513.1 | - | 7 | 691 | |||
BC130014.1 | - | 7 | 686 | 40140405 | ||
M64641.1 | - | 7 | 657 |