Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sik1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010831.2 | NP_034961.2 | 12 | 1869 | ||
GenBank mRNA | AK087322.1 | - | 5 | 786 | ||
AK141817.1 | - | 12 | 1869 | |||
AK172427.1 | - | 11 | 2309 | |||
AK220203.1 | - | 10 | 1482 | |||
U11494.2 | - | 12 | 1861 |