Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010833.2 | NP_034963.2 | 12 | 1578 | ||
GenBank mRNA | AK088336.1 | - | 12 | 1526 | ||
AK133819.1 | - | 12 | 1582 | |||
AK145746.1 | - | 12 | 1582 | |||
AK148721.1 | - | 2 | 81 | |||
AK149065.1 | - | 2 | 81 | |||
AK150869.1 | - | 12 | 1581 | |||
AK153210.1 | - | 12 | 1581 | |||
AK157701.1 | - | 12 | 1533 | |||
BC047366.1 | - | 12 | 1572 | 3711212 | ||
BQ950368.1 | - | 6 | 715 | |||
M86390.1 | - | 11 | 1358 | |||
S47577.1 | - | 12 | 1563 |