Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008634.2 | NP_032660.2 | 5 | 2580 | ||
GenBank mRNA | BC138033.1 | - | 5 | 2510 | 8861052 | |
BC138034.1 | - | 5 | 2510 | 8861053 | ||
X51396.1 | - | 5 | 2518 |