Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253877.1 | NP_001240806.1 | 7 | 1223 | ||
NM_001253878.1 | NP_001240807.1 | 7 | 942 | |||
NM_001253879.1 | NP_001240808.1 | 6 | 865 | |||
NM_001253880.1 | NP_001240809.1 | 4 | 580 | |||
NM_013827.3 | NP_038855.2 | 6 | 861 | |||
GenBank mRNA | AB221622.1 | - | 3 | 268 | ||
AK030752.1 | - | 6 | 811 | |||
AK031409.1 | - | 8 | 930 | |||
AK032798.1 | - | 7 | 1184 | |||
AK032819.1 | - | 7 | 1185 | |||
AK137539.1 | - | 8 | 1170 | |||
AK139685.1 | - | 7 | 943 | |||
BC050826.1 | - | 6 | 815 | |||
BC092237.1 | - | 6 | 793 | 30637248 | ||
BC100340.1 | - | 4 | 511 | 30715034 | ||
S78454.1 | - | 3 | 268 |