Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164190.1 | NP_001157662.1 | 8 | 813 | ||
NM_001164191.1 | NP_001157663.1 | 8 | 704 | |||
NM_001164192.1 | NP_001157664.1 | 8 | 704 | |||
NM_019926.3 | NP_064310.2 | 8 | 693 | |||
GenBank mRNA | AF073996.1 | - | 8 | 694 | ||
AK016445.1 | - | 8 | 814 | |||
AK031794.1 | - | 8 | 675 | |||
AK166452.1 | - | 8 | 672 | |||
AK171475.1 | - | 8 | 682 | |||
BC051022.1 | - | 8 | 662 | |||
BC066847.1 | - | 8 | 651 | |||
BC090984.1 | - | 8 | 671 | 30246206 |