Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mast2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042743.2 | NP_001036208.1 | 30 | 4630 | ||
NM_008641.3 | NP_032667.2 | 29 | 4612 | |||
GenBank mRNA | AB093264.1 | - | 20 | 3262 | ||
AK004728.1 | - | 11 | 2122 | |||
BC060703.1 | - | 30 | 4200 | 6412811 | ||
U02313.1 | - | 29 | 4282 |