Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144514.2 | NP_653097.2 | 2 | 457 | ||
GenBank mRNA | AB076722.1 | - | 3 | 1134 | ||
AK082909.1 | - | 2 | 434 | |||
AK133507.1 | - | 2 | 433 | |||
AK147766.1 | - | 295 | ||||
BC030052.1 | - | 2 | 357 | |||
BC038641.1 | - | 2 | 404 | |||
BC051210.1 | - | 2 | 418 | |||
BC132083.1 | - | 2 | 446 | 40130360 | ||
BC132085.1 | - | 2 | 447 | 40130362 | ||
D85430.1 | - | 2 | 427 |