Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF137598.1 | - | 2 | 160 | ||
AK009261.1 | - | 4 | 270 | |||
AK031084.1 | - | 2 | 905 | |||
AK149630.1 | - | 3 | 221 | |||
AK181570.1 | - | 1 | 14 | |||
AK187523.1 | - | 1 | 14 | |||
AK187805.1 | - | 1 | 14 | |||
BC005606.1 | - | 3 | 213 | 3498140 |