Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF137598.1 | - | 2 | 189 | ||
AK009261.1 | - | 4 | 299 | |||
AK031084.1 | - | 2 | 934 | |||
AK149630.1 | - | 3 | 250 | |||
AK181570.1 | - | 1 | 43 | |||
AK187523.1 | - | 1 | 43 | |||
AK187805.1 | - | 1 | 43 | |||
BC005606.1 | - | 3 | 242 | 3498140 |