Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162417.1 | NP_001155889.1 | 16, 17 | 1748 | ||
NM_001162418.1 | NP_001155890.1 | 14, 15 | 1625 | |||
NM_010852.2 | NP_034982.2 | 15, 16 | 1697 | |||
GenBank mRNA | AF483504.1 | - | 14, 15 | 1423 | ||
AF483505.1 | - | 14, 15 | 1423 | |||
AK012133.1 | - | 14, 15 | 1627 | |||
AK048404.1 | - | 16, 17 | 1715 | |||
AK129336.1 | - | 15, 16 | 1536 | |||
AK143230.1 | - | 15, 16 | 1633 |