Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053214.2 | NP_444444.2 | 15 | 1720 | ||
GenBank mRNA | AK036840.1 | - | 5 | 1553 | ||
AK131188.1 | - | 5 | 413 | |||
AK153908.1 | - | 15 | 1683 | |||
AK158333.1 | - | 15 | 1635 | |||
BC046502.1 | - | 15 | 1708 | 4167558 | ||
X97650.1 | - | 15 | 1638 |