Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256081.1 | NP_001243010.1 | 31 | 4298 | ||
NM_001256082.1 | NP_001243011.1 | 31 | 4145 | |||
NM_001256083.1 | NP_001243012.1 | 30 | 4127 | |||
NM_008663.2 | NP_032689.2 | 31 | 4298 | |||
GenBank mRNA | AY821853.1 | - | 30 | 4134 | ||
AY821854.1 | - | 31 | 4152 | |||
U81453.1 | - | 29 | 4048 |