Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083934.1 | NP_001077403.1 | 21 | 3116 | ||
NM_010867.2 | NP_034997.2 | 22 | 3410 | |||
GenBank mRNA | AJ012072.1 | - | 22 | 3443 | ||
AK041025.1 | - | 3 | 1399 | |||
BC060378.1 | - | 20 | 3021 | 4980452 | ||
Z22866.1 | - | 22 | 3389 |